siRNA Details
| Gene Symbol: Syne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764911.1 | XP_008763133.1 | 106 | 19674 | ||
| GenBank mRNA | BC098809.1 | - | 4 | 371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syne2| Gene Symbol: Syne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764911.1 | XP_008763133.1 | 106 | 19674 | ||
| GenBank mRNA | BC098809.1 | - | 4 | 371 | ||