siRNA Details
| Gene Symbol: Adck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108985.1 | NP_001102455.1 | 7 | 849 | ||
| XM_008764824.1 | XP_008763046.1 | 10 | 1064 | |||
| GenBank mRNA | FQ212472.1 | - | 8 | 1093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adck1| Gene Symbol: Adck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108985.1 | NP_001102455.1 | 7 | 849 | ||
| XM_008764824.1 | XP_008763046.1 | 10 | 1064 | |||
| GenBank mRNA | FQ212472.1 | - | 8 | 1093 | ||