siRNA Details
| Gene Symbol: Bin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012223.1 | NP_001012223.1 | 8 | 692 | ||
| XM_008765773.1 | XP_008763995.1 | 8 | 719 | |||
| GenBank mRNA | BC079406.1 | - | 8 | 692 | 7112822 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bin2| Gene Symbol: Bin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012223.1 | NP_001012223.1 | 8 | 692 | ||
| XM_008765773.1 | XP_008763995.1 | 8 | 719 | |||
| GenBank mRNA | BC079406.1 | - | 8 | 692 | 7112822 | |