siRNA Details
| Gene Symbol: Nfe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012224.1 | NP_001012224.1 | 2, 3 | 416 | ||
| XM_006242431.2 | XP_006242493.1 | 3, 4 | 628 | |||
| XM_006242432.2 | XP_006242494.1 | 2, 3 | 447 | |||
| XM_006242433.2 | XP_006242495.1 | 2, 3 | 1073 | |||
| GenBank mRNA | BC078925.1 | - | 2, 3 | 416 | 7106450 | |
| FQ233359.1 | - | 2, 3 | 250 | |||
| FQ234994.1 | - | 2, 3 | 261 | |||

Nfe2
Genomic Map