siRNA Details
| Gene Symbol: Zfp426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079943.1 | NP_001073412.1 | 8 | 710 | ||
| XM_006242666.1 | XP_006242728.1 | 7 | 717 | |||
| XM_006242667.2 | XP_006242729.1 | 7 | 2805 | |||
| XM_006242668.2 | XP_006242730.1 | 8 | 2111 | |||
| XM_006242670.1 | XP_006242732.1 | 8 | 800 | |||
| XM_006242671.2 | XP_006242733.1 | 6 | 586 | |||
| XM_006242672.2 | XP_006242734.1 | 7 | 2313 | |||
| XM_006242673.2 | XP_006242735.1 | 6 | 2166 | |||
| XM_008765981.1 | XP_008764203.1 | 7 | 2316 | |||
| XM_008765982.1 | XP_008764204.1 | 7 | 653 | |||
| XM_008765983.1 | XP_008764205.1 | 7 | 1961 | |||
| XM_008765984.1 | XP_008764206.1 | 7 | 1917 | |||
| GenBank mRNA | BC129125.1 | - | 8 | 710 | 7133611 | |

Zfp426
Genomic Map