siRNA Details
| Gene Symbol: Lrrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014268.1 | NP_001014290.1 | 7 | 722 | ||
| GenBank mRNA | BC079423.1 | - | 7 | 722 | 7114094 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc1| Gene Symbol: Lrrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014268.1 | NP_001014290.1 | 7 | 722 | ||
| GenBank mRNA | BC079423.1 | - | 7 | 722 | 7114094 | |