siRNA Details
Gene Symbol: Traip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109004.2 | NP_001102474.1 | 15 | 1389 | ||
GenBank mRNA | BC162006.1 | - | 15 | 1389 | 9089897 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Traip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109004.2 | NP_001102474.1 | 15 | 1389 | ||
GenBank mRNA | BC162006.1 | - | 15 | 1389 | 9089897 |