siRNA Details
| Gene Symbol: Ube2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008381.1 | NP_001008382.1 | 9 | 604 | ||
| XM_006245422.2 | XP_006245484.1 | 10 | 752 | |||
| XM_006245423.2 | XP_006245485.1 | 10 | 951 | |||
| XM_006245425.2 | XP_006245487.1 | 10 | 749 | |||
| XM_006245427.2 | XP_006245489.1 | 10 | 796 | |||
| XM_008767274.1 | XP_008765496.1 | 9 | 722 | |||
| XM_008767275.1 | XP_008765497.1 | 9 | 684 | |||
| GenBank mRNA | BC086355.1 | - | 9 | 604 | 7303709 | |

Ube2f
Genomic Map