siRNA Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015035.1 | NP_001015035.1 | 10, 11 | 1028 | ||
| XM_006247904.2 | XP_006247966.1 | 10, 11 | 1078 | |||
| XM_008768505.1 | XP_008766727.1 | 10, 11 | 1078 | |||
| XM_008768506.1 | XP_008766728.1 | 10, 11 | 1078 | |||
| XM_008768507.1 | XP_008766729.1 | 10, 11 | 1078 | |||
| XM_008768508.1 | XP_008766730.1 | 10, 11 | 1078 | |||
| GenBank mRNA | AY634365.1 | - | 10, 11 | 1028 | ||
| BC111711.1 | - | 11, 12 | 1077 | 7383845 | ||

B3gntl1
Genomic Map