siRNA Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199118.1 | NP_954549.1 | 3 | 799 | ||
| XM_006247905.1 | XP_006247967.1 | 3 | 779 | |||
| GenBank mRNA | BC061753.1 | - | 3 | 799 | 5599964 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gaa| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199118.1 | NP_954549.1 | 3 | 799 | ||
| XM_006247905.1 | XP_006247967.1 | 3 | 779 | |||
| GenBank mRNA | BC061753.1 | - | 3 | 799 | 5599964 | |