siRNA Details
| Gene Symbol: LOC102552805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301646.1 | NP_001288575.1 | 6 | 2451 | ||
| GenBank mRNA | AB275648.1 | - | 6 | 2451 | ||
| Gene Symbol: Ssx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773085.1 | XP_008771307.1 | 6 | 1721 | ||

LOC102552805
Genomic Map