siRNA Details
| Gene Symbol: Armcx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014274.1 | NP_001014296.1 | 5 | 2764 | ||
| XM_008773429.1 | XP_008771651.1 | 5 | 2874 | |||
| GenBank mRNA | BC081886.1 | - | 5 | 2764 | 7108660 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armcx2| Gene Symbol: Armcx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014274.1 | NP_001014296.1 | 5 | 2764 | ||
| XM_008773429.1 | XP_008771651.1 | 5 | 2874 | |||
| GenBank mRNA | BC081886.1 | - | 5 | 2764 | 7108660 | |