siRNA Details
Gene Symbol: Dgkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245504.2 | XP_006245566.2 | 9 | 992 | ||
XM_006245505.2 | XP_006245567.2 | 9 | 992 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dgkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245504.2 | XP_006245566.2 | 9 | 992 | ||
XM_006245505.2 | XP_006245567.2 | 9 | 992 |