siRNA Details
| Gene Symbol: Pigw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194461.1 | NP_919443.1 | 1 | 210 | ||
| XM_006247055.2 | XP_006247117.1 | 2 | 375 | |||
| XM_006247056.2 | XP_006247118.1 | 2 | 298 | |||
| XM_006247057.2 | XP_006247119.1 | 2 | 280 | |||

Pigw
Genomic Map