siRNA Details
| Gene Symbol: Iqsec3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207617.1 | NP_997500.1 | 8 | 3216 | ||
| GenBank mRNA | AB057643.1 | - | 8 | 3216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqsec3| Gene Symbol: Iqsec3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207617.1 | NP_997500.1 | 8 | 3216 | ||
| GenBank mRNA | AB057643.1 | - | 8 | 3216 | ||