siRNA Details
Gene Symbol: Olr1404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000787.1 | NP_001000787.1 | 1 | 543 | ||
GenBank mRNA | X89699.1 | - | 1 | 169 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000787.1 | NP_001000787.1 | 1 | 543 | ||
GenBank mRNA | X89699.1 | - | 1 | 169 |