siRNA Details
| Gene Symbol: Olr1331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000790.1 | NP_001000790.1 | 761 | |||
| XM_008766124.1 | XP_008764346.1 | 2 | 650 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1331| Gene Symbol: Olr1331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000790.1 | NP_001000790.1 | 761 | |||
| XM_008766124.1 | XP_008764346.1 | 2 | 650 | |||