siRNA Details
Gene Symbol: Olr804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000852.1 | NP_001000852.1 | 1 | 588 | ||
XM_008762900.1 | XP_008761122.1 | 2 | 702 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000852.1 | NP_001000852.1 | 1 | 588 | ||
XM_008762900.1 | XP_008761122.1 | 2 | 702 |