siRNA Details
| Gene Symbol: Olr1191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000981.1 | NP_001000981.1 | 1 | 139 | ||
| Gene Symbol: LOC100361194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729873.3 | XP_002729919.2 | 1 | 139 | ||

Olr1191
Genomic Map