siRNA Details
| Gene Symbol: Olr445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001049.1 | NP_001001049.1 | 1 | 295 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr445| Gene Symbol: Olr445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001049.1 | NP_001001049.1 | 1 | 295 | ||