siRNA Details
| Gene Symbol: Fkbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002818.2 | NP_001002818.1 | 2 | 217 | ||
| GenBank mRNA | BC086532.1 | - | 2 | 217 | 7305083 | |
| FQ231484.1 | - | 2 | 231 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbpl| Gene Symbol: Fkbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002818.2 | NP_001002818.1 | 2 | 217 | ||
| GenBank mRNA | BC086532.1 | - | 2 | 217 | 7305083 | |
| FQ231484.1 | - | 2 | 231 | |||