siRNA Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002822.2 | NP_001002822.2 | 2102 | |||
| GenBank mRNA | BC091220.1 | - | 10 | 1560 | ||
| BC105619.1 | - | 2100 | 7453057 | |||
| FQ225184.1 | - | 3 | 224 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cchcr1| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002822.2 | NP_001002822.2 | 2102 | |||
| GenBank mRNA | BC091220.1 | - | 10 | 1560 | ||
| BC105619.1 | - | 2100 | 7453057 | |||
| FQ225184.1 | - | 3 | 224 | |||