siRNA Details
| Gene Symbol: Cpa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002808.2 | NP_001002808.2 | 5 | 873 | ||
| XM_006236215.2 | XP_006236277.1 | 6 | 1412 | |||
| XM_006236216.2 | XP_006236278.1 | 6 | 1136 | |||
| XM_006236217.2 | XP_006236279.1 | 6 | 1096 | |||
| XM_006236218.2 | XP_006236280.1 | 6 | 667 | |||
| XM_006236219.2 | XP_006236281.1 | 5 | 1561 | |||
| XM_008762801.1 | XP_008761023.1 | 6 | 1452 | |||
| XM_008762802.1 | XP_008761024.1 | 6 | 1428 | |||
| XM_008762803.1 | XP_008761025.1 | 5 | 1561 | |||
| GenBank mRNA | BC085840.1 | - | 5 | 873 | 7135802 | |

Cpa5
Genomic Map