siRNA Details
| Gene Symbol: Sgce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002023.2 | NP_001002023.1 | 8 | 1269 | ||
| GenBank mRNA | AY164274.1 | - | 8 | 1230 | ||
| BC089947.1 | - | 8 | 1269 | 7311584 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgce| Gene Symbol: Sgce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002023.2 | NP_001002023.1 | 8 | 1269 | ||
| GenBank mRNA | AY164274.1 | - | 8 | 1230 | ||
| BC089947.1 | - | 8 | 1269 | 7311584 | ||