siRNA Details
| Gene Symbol: C5ar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003710.2 | NP_001003710.2 | 2 | 658 | ||
| XM_006228352.2 | XP_006228414.1 | 2 | 1040 | |||
| XM_006228353.2 | XP_006228415.1 | 2 | 812 | |||
| GenBank mRNA | AY600435.1 | - | 1 | 524 | ||
| AY741660.1 | - | 2 | 658 | |||
More Information
Set Membership: |
|

C5ar2
Genomic Map