siRNA Details
| Gene Symbol: Kcnk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003820.1 | NP_001003820.1 | 2, 3 | 408 | ||
| GenBank mRNA | AY567970.1 | - | 2, 3 | 408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk18| Gene Symbol: Kcnk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003820.1 | NP_001003820.1 | 2, 3 | 408 | ||
| GenBank mRNA | AY567970.1 | - | 2, 3 | 408 | ||