siRNA Details
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031606.1 | NP_113794.1 | 6 | 620 | ||
GenBank mRNA | AF017185.1 | - | 6 | 620 | ||
AF455569.1 | - | 6 | 620 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031606.1 | NP_113794.1 | 6 | 620 | ||
GenBank mRNA | AF017185.1 | - | 6 | 620 | ||
AF455569.1 | - | 6 | 620 |