siRNA Details
| Gene Symbol: Npepps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080395.1 | NP_536320.1 | 20 | 2466 | ||
| GenBank mRNA | CA503636.1 | - | 4 | 600 | ||
| DY316730.1 | - | 5 | 605 | |||
| FQ209839.1 | - | 15 | 1573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npepps| Gene Symbol: Npepps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080395.1 | NP_536320.1 | 20 | 2466 | ||
| GenBank mRNA | CA503636.1 | - | 4 | 600 | ||
| DY316730.1 | - | 5 | 605 | |||
| FQ209839.1 | - | 15 | 1573 | |||