siRNA Details
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017111.1 | NP_058807.1 | 8, 9 | 994 | ||
GenBank mRNA | FQ209854.1 | - | 8, 9 | 1012 | ||
L19031.1 | - | 8, 9 | 994 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017111.1 | NP_058807.1 | 8, 9 | 994 | ||
GenBank mRNA | FQ209854.1 | - | 8, 9 | 1012 | ||
L19031.1 | - | 8, 9 | 994 |