siRNA Details
Gene Symbol: Bpifa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052808.1 | NP_434695.1 | 2 | 61 | ||
XM_008762379.1 | XP_008760601.1 | 3 | 410 | |||
GenBank mRNA | M83209.1 | - | 2 | 61 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bpifa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052808.1 | NP_434695.1 | 2 | 61 | ||
XM_008762379.1 | XP_008760601.1 | 3 | 410 | |||
GenBank mRNA | M83209.1 | - | 2 | 61 |