siRNA Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017317.2 | NP_059013.1 | 2 | 136 | ||
| GenBank mRNA | BC098667.1 | - | 2 | 136 | 7384913 | |
| D17352.1 | - | 1 | 72 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab27a| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017317.2 | NP_059013.1 | 2 | 136 | ||
| GenBank mRNA | BC098667.1 | - | 2 | 136 | 7384913 | |
| D17352.1 | - | 1 | 72 | |||