siRNA Details
| Gene Symbol: Ptk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017318.2 | NP_059014.2 | 21 | 1949 | ||
| XM_006252143.2 | XP_006252205.1 | 21 | 2274 | |||
| XM_006252144.2 | XP_006252206.1 | 21 | 5265 | |||
| XM_006252145.2 | XP_006252207.1 | 21 | 2078 | |||
| XM_008770773.1 | XP_008768995.1 | 21 | 3462 | |||
| XM_008770774.1 | XP_008768996.1 | 21 | 3028 | |||
| GenBank mRNA | BC101921.1 | - | 21 | 1949 | 7937929 | |
| D45854.1 | - | 21 | 2127 | |||
| U69109.1 | - | 20 | 1869 | |||

Ptk2b
Genomic Map