siRNA Details
| Gene Symbol: Aco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017321.1 | NP_059017.1 | 19, 20 | 2597 | ||
| GenBank mRNA | FQ213654.1 | - | 4, 5 | 428 | ||
| L23874.1 | - | 19, 20 | 2597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aco1| Gene Symbol: Aco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017321.1 | NP_059017.1 | 19, 20 | 2597 | ||
| GenBank mRNA | FQ213654.1 | - | 4, 5 | 428 | ||
| L23874.1 | - | 19, 20 | 2597 | |||