siRNA Details
| Gene Symbol: Mapk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270544.1 | NP_001257473.1 | 10 | 1296 | ||
| NM_001270545.1 | NP_001257474.1 | 10 | 1296 | |||
| NM_017322.2 | NP_059018.1 | 10 | 1296 | |||
| XM_006246317.1 | XP_006246379.1 | 10 | 1292 | |||
| XM_006246318.2 | XP_006246380.1 | 10 | 1244 | |||
| GenBank mRNA | BC061870.1 | - | 10 | 1263 | 5598943 | |
| L27111.1 | - | 9 | 1052 | |||
| L27112.1 | - | 9 | 1052 | |||

Mapk9
Genomic Map