siRNA Details
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270544.1 | NP_001257473.1 | 11, 12 | 1387 | ||
NM_017322.2 | NP_059018.1 | 11, 12 | 1387 | |||
XM_006246317.1 | XP_006246379.1 | 11, 12 | 1383 | |||
XM_006246318.2 | XP_006246380.1 | 11, 12 | 1335 | |||
GenBank mRNA | FQ234999.1 | - | 1, 2 | 43 | ||
L27111.1 | - | 10, 11 | 1143 | |||
L27112.1 | - | 10, 11 | 1143 |
