siRNA Details
| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017323.1 | NP_059019.1 | 7 | 848 | ||
| GenBank mRNA | L27513.1 | - | 7 | 848 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr2c2| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017323.1 | NP_059019.1 | 7 | 848 | ||
| GenBank mRNA | L27513.1 | - | 7 | 848 | ||