siRNA Details
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017327.1 | NP_059023.1 | 8 | 1698 | ||
| XM_008772320.1 | XP_008770542.1 | 8 | 1707 | |||
| XM_008772322.1 | XP_008770544.1 | 7 | 4316 | |||
| GenBank mRNA | M12671.1 | - | 7 | 862 | ||
| M17526.1 | - | 8 | 1698 | |||

Gnao1
Genomic Map