siRNA Details
| Gene Symbol: Ptpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017336.1 | NP_059032.1 | 24 | 3446 | ||
| XM_006237571.2 | XP_006237633.1 | 13 | 1354 | |||
| XM_006237572.2 | XP_006237634.1 | 12 | 1268 | |||
| GenBank mRNA | D30749.1 | - | 2 | 249 | ||
| D45412.1 | - | 15 | 1685 | |||
| U28938.1 | - | 24 | 3446 | |||

Ptpro
Genomic Map