siRNA Details
| Gene Symbol: Pde3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017337.1 | NP_059033.1 | 13 | 2649 | ||
| XM_008763422.1 | XP_008761644.1 | 13 | 2717 | |||
| GenBank mRNA | U38179.1 | - | 13 | 2649 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde3a| Gene Symbol: Pde3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017337.1 | NP_059033.1 | 13 | 2649 | ||
| XM_008763422.1 | XP_008761644.1 | 13 | 2717 | |||
| GenBank mRNA | U38179.1 | - | 13 | 2649 | ||