siRNA Details
| Gene Symbol: Gsk3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017344.1 | NP_059040.1 | 4 | 765 | ||
| XM_008758972.1 | XP_008757194.1 | 4 | 774 | |||
| GenBank mRNA | X53427.1 | - | 4 | 765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsk3a| Gene Symbol: Gsk3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017344.1 | NP_059040.1 | 4 | 765 | ||
| XM_008758972.1 | XP_008757194.1 | 4 | 774 | |||
| GenBank mRNA | X53427.1 | - | 4 | 765 | ||