siRNA Details
| Gene Symbol: Cacnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017346.1 | NP_059042.1 | 11, 12 | 1119 | ||
| XM_006247409.2 | XP_006247471.1 | 11, 12 | 1387 | |||
| XM_006247410.2 | XP_006247472.1 | 10, 11 | 2329 | |||
| XM_006247411.2 | XP_006247473.1 | 10, 11 | 2329 | |||
| GenBank mRNA | X61394.1 | - | 11, 12 | 1119 | ||

Cacnb1
Genomic Map