siRNA Details
| Gene Symbol: Slc6a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017348.2 | NP_059044.1 | 10, 11 | 2092 | ||
| XM_006229611.1 | XP_006229673.1 | 9, 10 | 1989 | |||
| GenBank mRNA | BC168238.1 | - | 10, 11 | 2070 | 5598758 | |

Slc6a8
Genomic Map