siRNA Details
| Gene Symbol: Plaur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017350.1 | NP_059046.1 | 4 | 442 | ||
| NM_134352.1 | NP_599179.1 | 4 | 447 | |||
| GenBank mRNA | AF007789.1 | - | 4 | 434 | ||
| BC127499.1 | - | 4 | 442 | 7380245 | ||
| FQ220340.1 | - | 4 | 455 | |||
| X71898.1 | - | 4 | 447 | |||
| X71899.1 | - | 4 | 442 | |||

Plaur
Genomic Map