siRNA Details
| Gene Symbol: Nup54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017361.1 | NP_059057.1 | 6, 7 | 986 | ||
| GenBank mRNA | BC078858.1 | - | 6, 7 | 1016 | 7134682 | |
| U63840.1 | - | 6, 7 | 986 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup54| Gene Symbol: Nup54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017361.1 | NP_059057.1 | 6, 7 | 986 | ||
| GenBank mRNA | BC078858.1 | - | 6, 7 | 1016 | 7134682 | |
| U63840.1 | - | 6, 7 | 986 | |||