siRNA Details
| Gene Symbol: Chrm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017362.4 | NP_059058.1 | 2113 | |||
| GenBank mRNA | M22926.1 | - | 2113 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_205783, BC120615.
Set Membership: |
|

Chrm5
Genomic Map