siRNA Details
| Gene Symbol: Cltc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019299.1 | NP_062172.1 | 18 | 2976 | ||
| XM_006247107.2 | XP_006247169.1 | 19 | 3288 | |||
| XM_006247108.2 | XP_006247170.1 | 18 | 3284 | |||
| XM_006247109.2 | XP_006247171.1 | 19 | 3287 | |||
| XM_008768108.1 | XP_008766330.1 | 17 | 3503 | |||
| GenBank mRNA | J03583.1 | - | 18 | 2976 | ||

Cltc
Genomic Map