siRNA Details
| Gene Symbol: Dgkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019304.1 | NP_062177.1 | 15 | 1531 | ||
| XM_008764649.1 | XP_008762871.1 | 14 | 1478 | |||
| XM_008764650.1 | XP_008762872.1 | 14 | 1475 | |||
| XM_008764651.1 | XP_008762873.1 | 13 | 1452 | |||
| GenBank mRNA | D16100.1 | - | 15 | 1531 | ||

Dgkb
Genomic Map