siRNA Details
| Gene Symbol: Fgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019305.2 | NP_062178.1 | 2 | 776 | ||
| GenBank mRNA | M22427.1 | - | 776 | |||
| X07285.1 | - | 2 | 497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf2| Gene Symbol: Fgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019305.2 | NP_062178.1 | 2 | 776 | ||
| GenBank mRNA | M22427.1 | - | 776 | |||
| X07285.1 | - | 2 | 497 | |||