siRNA Details
| Gene Symbol: Kcnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019313.1 | NP_062186.1 | 3 | 696 | ||
| XM_006252933.2 | XP_006252995.1 | 3 | 729 | |||
| GenBank mRNA | AF000973.1 | - | 3 | 696 | ||
| U69885.1 | - | 3 | 462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnn1| Gene Symbol: Kcnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019313.1 | NP_062186.1 | 3 | 696 | ||
| XM_006252933.2 | XP_006252995.1 | 3 | 729 | |||
| GenBank mRNA | AF000973.1 | - | 3 | 696 | ||
| U69885.1 | - | 3 | 462 | |||