siRNA Details
| Gene Symbol: Kcnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019315.2 | NP_062188.2 | 1 | 684 | ||
| GenBank mRNA | AF284345.1 | - | 1 | 381 | ||
| AF292389.1 | - | 1 | 684 | |||
| U69884.1 | - | 1 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnn3| Gene Symbol: Kcnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019315.2 | NP_062188.2 | 1 | 684 | ||
| GenBank mRNA | AF284345.1 | - | 1 | 381 | ||
| AF292389.1 | - | 1 | 684 | |||
| U69884.1 | - | 1 | 388 | |||